Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-2,3-O-Benzylidene-D-threitol, 98%, Thermo Scientific™

Catalog No. AC254840010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 58383-35-0
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.3
MDL Number MFCD00010198
InChI Key AEJRVTSEJAYBNR-NXEZZACHSA-N
Synonym +-2,3-o-benzylidene-d-threitol, 4r,5r-2-phenyl-1,3-dioxolane-4,5-diyl dimethanol, 4r,5r-5-hydroxymethyl-2-phenyl-1,3-dioxolan-4-yl methanol, 2,3-o-benzylidene-d-threitol, 2-o,3-o-benzylidene-d-threitol, --2,3-0-benzylidene-l-threitol, 4r,5r-4,5-dihydroxymethyl-2-phenyl-1,3-dioxolan, 4r,5r-+-2-phenyl-1,3-dioxolane-4,5-dimethanol, 4r,5r-5-hydroxymethyl-2-phenyl-1,3-dioxolan-4-yl methan-1-ol
PubChem CID 719729
IUPAC Name [(4R,5R)-5-(hydroxymethyl)-2-phenyl-1,3-dioxolan-4-yl]methanol
SMILES C1=CC=C(C=C1)C2OC(C(O2)CO)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 69.0°C to 74.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation + 10.50
Formula Weight 210.3
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (+)-2, 3-O-Benzylidene-D-threitol, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.