Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trimethyloxazole 98.0+%, TCI America™
SDP

Catalog No. t25955g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 20662-84-4
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.144
MDL Number MFCD00005308
InChI Key ZRLDBDZSLLGDOX-UHFFFAOYSA-N
Synonym 2,4,5-trimethyloxazole, trimethyloxazole, oxazole, trimethyl, unii-b04pf51wxi, 2,4,5-trimethyl oxazole, trimethyl-1,3-oxazole, b04pf51wxi, oxazole, 2,4,5-trimethyl, trimethyl-oxazole, pubchem8636
PubChem CID 30215
IUPAC Name 2,4,5-trimethyl-1,3-oxazole
SMILES CC1=C(OC(=N1)C)C

Specifications

Color Yellow
Boiling Point 134°C
Quantity 5 g
UN Number 1993
Formula Weight 111.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trimethyloxazole
EINECSNumber (5)-3394
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.