Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tri(dimethylaminomethyl)phenol, MP Biomedicals

Catalog No. icn219126 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g

Chemical Identifiers

CAS 90-72-2
Molecular Formula C15H27N3O
Molecular Weight (g/mol) 265.401
InChI Key AHDSRXYHVZECER-UHFFFAOYSA-N
Synonym 2,4,6-tris dimethylaminomethyl phenol, sumicure d, 2,4,6-tris dimethylamino methyl phenol, dmf 3, actiron nx 3, ancamine k 54, capcure eh 30, epilink 230, araldite dy 061
PubChem CID 7026
IUPAC Name 2,4,6-tris[(dimethylamino)methyl]phenol
SMILES CN(C)CC1=CC(=C(C(=C1)CN(C)C)O)CN(C)C

Specifications

Quantity 100 g
Formula Weight 265.4
Chemical Name or Material 2, 4, 6-Tri(Dimethylaminomethyl)Phenol
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.