Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,4,6-Triphenylaniline, 98%

Click to view available options
Quantity:
1g
5g
25g
6864-20-6
C24H19N
321.423
MFCD00012371
AGWKGKUGJDMKMT-UHFFFAOYSA-N
2,4,6-triphenyl-anilin,2,4,6-triphenylphenylamine,bidd:gt0713,2,4,6-triphenylaniline,3,5-diphenyl-1,1'-biphenyl-2-amine,1,1':3',1-terphenyl-2'-amine,5'-phenyl,5'-phenyl-1,1':3',1-terphenyl-2'-amine,1,1':3',1-terphenyl-2'-amine, 5'-phenyl
629425
2,4,6-triphenylaniline
C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4
6864-20-6
C24H19N
321.423
MFCD00012371
AGWKGKUGJDMKMT-UHFFFAOYSA-N
2,4,6-triphenyl-anilin,2,4,6-triphenylphenylamine,bidd:gt0713,2,4,6-triphenylaniline,3,5-diphenyl-1,1'-biphenyl-2-amine,1,1':3',1-terphenyl-2'-amine,5'-phenyl,5'-phenyl-1,1':3',1-terphenyl-2'-amine,1,1':3',1-terphenyl-2'-amine, 5'-phenyl
629425
2,4,6-triphenylaniline
C1=CC=C(C=C1)C2=CC(=C(C(=C2)C3=CC=CC=C3)N)C4=CC=CC=C4
Melting Point 122°C to 125°C
Quantity 25g
Formula Weight 321.43
Percent Purity 98%
Chemical Name or Material 2,4,6-Triphenylaniline

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger