Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Bromobutyl)-1,3-dioxolane 95.0+%, TCI America™
SDP

Catalog No. B212825G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 87227-41-6
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.083
MDL Number MFCD02093454
InChI Key UQTZEBQZCBQLAN-UHFFFAOYSA-N
Synonym 5-Bromovaleraldehyde Ethylene Acetal
PubChem CID 13780200
IUPAC Name 2-(4-bromobutyl)-1,3-dioxolane
SMILES C1COC(O1)CCCCBr

Specifications

Color Yellow
Quantity 25 g
Formula Weight 209.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Bromobutyl)-1,3-dioxolane
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.