Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluoro-1-nitrobenzene, 98%

Catalog No. AAA1324922
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1324922 100 g
AAA1324914 25 g
AAA1324936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1324922 Supplier Thermo Scientific Chemicals Supplier No. A1324922
Only null left
Add to Cart

CAS: 446-35-5 | C6H3F2NO2 | 159.09 g/mol

2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoronitrobenzene has been used in the synthesis of 2,4-difluoro-5-nitrobenzenesulfonic acid via sulfonation reaction, (±)-horsfiline and resin-bound-2-arylaminobenzimidazoles.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-35-5
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00007050
InChI Key RJXOVESYJFXCGI-UHFFFAOYSA-N
Synonym 2,4-difluoronitrobenzene, benzene, 2,4-difluoro-1-nitro, 1-nitro-2,4-difluorobenzene, 1,3-difluoro-4-nitrobenzene, unii-6398y1d7js, 2,4-difluoro-1-nitro-benzene, pubchem2288, 2,4-difluornitrobenzol, 2,4-difluoronitrobenzol, acmc-1akft
PubChem CID 67967
IUPAC Name 2,4-difluoro-1-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(F)C=C1F

Specifications

Melting Point 9°C to 10°C
Density 1.45
Boiling Point 203°C to 204°C
Flash Point 90°C (194°F)
Refractive Index 1.513
Quantity 100 g
UN Number UN2810
Beilstein 2048034
Solubility Information Insoluble in water.
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 2,4-Difluoro-1-nitrobenzene
Show More Show Less
Hazard Category H227-H301+H311+H331-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H331-H373-H227-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 207-167-0
RTECSNumber CZ5730000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.