Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluoroiodobenzene, 99+%, Thermo Scientific™

Catalog No. AC269780100
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2265-93-2
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD00040151
InChI Key YKLDMAPEGQYZRT-UHFFFAOYSA-N
Synonym 2,4-difluoroiodobenzene, 1,3-difluoro-4-iodobenzene, 2,4-difluoro-1-iodobenze, benzene, 2,4-difluoro-1-iodo, 2,4-difluoro iodobenzene, 2,4-difluoro-1-iodo-benzene, pubchem3463, 2,4-difluoro-iodobenzene, acmc-1cq0v, intermediates-zcf02607
PubChem CID 137519
IUPAC Name 2,4-difluoro-1-iodobenzene
SMILES FC1=CC=C(I)C(F)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Brown to Yellow
Density 2.0000g/mL
Boiling Point 175°C to 176°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.5570 to 1.559
Quantity 10 g
Specific Gravity 2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 239.98
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2, 4-Difluoroiodobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.