Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dihydroxy-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. d40025g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 89282-12-2
Molecular Formula C5H4N2O4
Molecular Weight (g/mol) 156.097
MDL Number MFCD01075671
InChI Key BKYGVGWYPFVKTK-UHFFFAOYSA-N
Synonym 2,4-dihydroxy-3-nitropyridine, 3-nitropyridine-2,4-diol, 3-nitro-2,4-pyridinediol, 4-hydroxy-3-nitro-2-pyridone, 2-hydroxy-3-nitro-1h-pyridin-4-one, 2 1h-pyridinone, 4-hydroxy-3-nitro, pubchem5469, acmc-209qzb, 3-nitro-pyridine-2,4-diol, ksc495s9n
PubChem CID 54676840
IUPAC Name 4-hydroxy-3-nitro-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1O)[N+](=O)[O-]

Specifications

Melting Point 264°C
Quantity 5 g
Formula Weight 156.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dihydroxy-3-nitropyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.