Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. D232725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D232725G 25 g
D23275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D232725G Supplier TCI America Supplier No. D232725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3551-55-1
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.142
MDL Number MFCD00056028
InChI Key KEVRHVMWBKFGLO-UHFFFAOYSA-N
Synonym pyrimidine, 2,4-dimethoxy, unii-980k6dh82b, dimethoxypyrimidine, zlchem 892, pubchem7104, acmc-1adzt, 2,4-dimethoxy-pyrimidine, ksc911k2l, 2,4-dimethoxypyrimidine, kevrhvmwbkfglo-uhfffaoysa
PubChem CID 137976
IUPAC Name 2,4-dimethoxypyrimidine
SMILES COC1=NC(=NC=C1)OC

Specifications

Color Colorless
Boiling Point 205°C
Quantity 25 g
Formula Weight 140.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.