Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethyl-5-phenylthiophene 98.0+%, TCI America™
SDP

Catalog No. d32281g
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 57021-49-5
Molecular Formula C12H12S
Molecular Weight (g/mol) 188.288
InChI Key RLISGGFOYMIIRQ-UHFFFAOYSA-N
PubChem CID 10607839
IUPAC Name 3,5-dimethyl-2-phenylthiophene
SMILES CC1=CC(=C(S1)C2=CC=CC=C2)C

Specifications

Color Yellow
Boiling Point 151°C
Quantity 1 g
Formula Weight 188.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethyl-5-phenylthiophene
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.