Learn More
2,5-Diaminohydroquinone dihydrochloride, 97%
Description
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.
Notes
Air sensitive. It may discolor to black-violet on storage. Store away from strong oxidizing agents, air.
Specifications
Specifications
| Melting Point | >300°C |
| Odor | Odorless |
| Quantity | 1 g |
| Sensitivity | Air Sensitive |
| Formula Weight | 213.06 |
| Percent Purity | 97% |
| Chemical Name or Material | 2,5-Diaminohydroquinone dihydrochloride |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.