Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorophenyl Isocyanate 97.0+%, TCI America™
SDP

Catalog No. D03975G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. D03975G

Supplier: TCI America D03975G

Chemical Identifiers

CAS 5392-82-5
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002002
InChI Key PEQMJVGRHNZPAM-UHFFFAOYSA-N
Synonym 2,5-dichlorophenyl isocyanate, benzene, 1,4-dichloro-2-isocyanato, 2,5-dichlorophenylisocyanate, isocyanic acid, 2,5-dichlorophenyl ester, isocyanic acid 2,5-dichlorophenyl ester, acmc-209laz, 2,5-dichlorobenzenisocyanate, benzene,4-dichloro-2-isocyanato, #, isocyanic acid,5-dichlorophenyl ester
PubChem CID 79360
IUPAC Name 1,4-dichloro-2-isocyanatobenzene
SMILES ClC1=CC=C(Cl)C(=C1)N=C=O

Specifications

Color White-Yellow
Boiling Point 126°C
Quantity 5 g
UN Number 2250
Formula Weight 188.01
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 2,5-Dichlorophenyl Isocyanate
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.