Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylpyrrole 98.0+%, TCI America™
SDP

Catalog No. D026025ML Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 625-84-3
Molecular Formula C6H9N
Molecular Weight (g/mol) 95.145
MDL Number MFCD00005223
InChI Key PAPNRQCYSFBWDI-UHFFFAOYSA-N
Synonym 2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole
PubChem CID 12265
IUPAC Name 2,5-dimethyl-1H-pyrrole
SMILES CC1=CC=C(N1)C

Specifications

Color Yellow
Boiling Point 169°C
Quantity 25 mL
UN Number 2929
Formula Weight 95.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylpyrrole
RTECSNumber UX9465930
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.