Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[(5-Nitro-2-pyridyl)oxy]ethan-1-ol, 97%, Thermo Scientific™

Catalog No. p-2697500
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle

Catalog No. CD08075DA

Supplier: Thermo Scientific Chemicals CD08075DA

Only null left
Add to Cart

Chemical Identifiers

CAS 143071-39-0
Molecular Formula C7H8N2O4
Molecular Weight (g/mol) 184.151
MDL Number MFCD00052642
InChI Key KESQFSZFUCZCEI-UHFFFAOYSA-N
Synonym 2-5-nitropyridin-2-yloxy ethanol, 2-5-nitro-2-pyridyloxy ethanol, 2-5-nitropyridin-2-yl oxy ethan-1-ol, 2-5-nitro-2-pyridyl oxy ethan-1-ol, 2-2-hydroxyethoxy-5-nitropyridine, 2-5-nitropyridin-2-yl oxy ethanol, 2-5-nitro-2-pyridyloxy ethan-1-ol, acmc-209xhf, maybridge1_002682, 2-5-nitropyridin-2-yl oxyethanol
PubChem CID 587937
IUPAC Name 2-(5-nitropyridin-2-yl)oxyethanol
SMILES C1=CC(=NC=C1[N+](=O)[O-])OCCO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.15
Percent Purity 97%
Chemical Name or Material 2-[(5-Nitro-2-pyridyl)oxy]ethan-1-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.