Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichlorobenzylamine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
6575-27-5
C7H7Cl2N
176.05
MFCD00047928
VLVLNNQMURDGPM-UHFFFAOYSA-N
2,6-dichlorobenzylamine,2,6-dichlorophenyl methanamine,benzenemethanamine, 2,6-dichloro,2,6-dichlorophenyl methylamine,1-2,6-dichlorophenyl methanamine,pubchem15690,2,6-dichloro-benzylamine,acmc-20ao98,chembl13305,benzenemethanamine,2,6-dichloro
485432
(2,6-dichlorophenyl)methanamine
C1=CC(=C(C(=C1)Cl)CN)Cl
6575-27-5
C7H7Cl2N
176.05
MFCD00047928
VLVLNNQMURDGPM-UHFFFAOYSA-N
2,6-dichlorobenzylamine,2,6-dichlorophenyl methanamine,benzenemethanamine, 2,6-dichloro,2,6-dichlorophenyl methylamine,1-2,6-dichlorophenyl methanamine,pubchem15690,2,6-dichloro-benzylamine,acmc-20ao98,chembl13305,benzenemethanamine,2,6-dichloro
485432
(2,6-dichlorophenyl)methanamine
C1=CC(=C(C(=C1)Cl)CN)Cl
Refractive Index 1.5570 to 1.559
Packaging Glass bottle
Color Colorless to Yellow
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 12, III, 2347
Formula Weight 176.05
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 2, 6-Dichlorobenzylamine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger