Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-p-benzoquinone, 99%

Catalog No. AAA1134203
Click to view available options
Quantity:
1 g
5 g
25 g

Catalog No. AAA1134203

Supplier: Thermo Scientific Chemicals A1134203

Only null left
Add to Cart

CAS: 527-61-7 | C8H8O2 | 136.15 g/mol

2,6-Dimethyl-p-benzoquinone is used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dimethyl-p-benzoquinone is used as organic intermediates.

Solubility
Soluble in chloroform. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527-61-7
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00001605
InChI Key SENUUPBBLQWHMF-UHFFFAOYSA-N
Synonym 2,6-dimethylbenzoquinone, 2,6-dimethyl-1,4-benzoquinone, 2,6-dimethyl-p-benzoquinone, m-xyloquinone, 2,6-xyloquinone, 2,6-dimethylquinone, 2,5-cyclohexadiene-1,4-dione, 2,6-dimethyl, 3,5-dimethylbenzoquinone, p-benzoquinone, 2,6-dimethyl, 2,6-dimethyl-1,4-quinone
PubChem CID 68241
IUPAC Name 2,6-dimethylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C=C(C1=O)C

Specifications

Melting Point 69°C to 71°C
Quantity 1 g
Beilstein 2041345
Solubility Information Soluble in chloroform. Insoluble in water.
Formula Weight 136.15
Percent Purity 99%
Chemical Name or Material 2,6-Dimethyl-p-benzoquinone
Hazard Category H315-H319-H335-H500
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-420-8
RTECSNumber DK4825000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.