Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene 98.0+%, TCI America™
SDP

Catalog No. B49261G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Catalog No. B49261G

Supplier: TCI America B49261G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 222319-05-3
Molecular Formula C47H36N2
Molecular Weight (g/mol) 628.82
MDL Number MFCD11110702
InChI Key KJEQVQJWXVHKGT-UHFFFAOYSA-N
Synonym 9,9-Dimethyl-N,N′C-di(1-naphthyl)-N,N′C-diphenyl-9H-fluorene-2,7-diamine, DMFL-NPB
PubChem CID 20745597
IUPAC Name 9,9-dimethyl-N2,N7-bis(naphthalen-1-yl)-N2,N7-diphenyl-9H-fluorene-2,7-diamine
SMILES CC1(C)C2=C(C=CC(=C2)N(C2=CC=CC=C2)C2=C3C=CC=CC3=CC=C2)C2=C1C=C(C=C2)N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 267°C
Color White-Yellow
Quantity 1 g
Formula Weight 628.82
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.