Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylthiazole, 99%

Catalog No. p-7081335
Click to view available options
Quantity:
1 g
5 g

CAS: 24295-03-2 | C5H5NOS | 127.161 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24295-03-2
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.161
MDL Number MFCD00005324
InChI Key MOMFXATYAINJML-UHFFFAOYSA-N
Synonym 2-acetylthiazole, 1-1,3-thiazol-2-yl ethan-1-one, ethanone, 1-2-thiazolyl, 1-1,3-thiazol-2-yl ethanone, 2-acetyl thiazole, 1-2-thiazolyl ethanone, ketone, methyl 2-thiazolyl, 2-acetylthiazol, 2-acetyl-1,3-thiazole, 1-thiazol-2-yl-ethanone
PubChem CID 520108
IUPAC Name 1-(1,3-thiazol-2-yl)ethanone
SMILES CC(=O)C1=NC=CS1

Specifications

Density 1.227
Boiling Point 89°C to 91°C (12 mmHg)
Flash Point 78°C (172°F)
Odor Characteristic
Refractive Index 1.548
Quantity 1 g
Beilstein 109803
Formula Weight 127.17
Percent Purity 99%
Chemical Name or Material 2-Acetylthiazole
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 246-134-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.