Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Allyloxyethanol, 98%

Catalog No. AC449032500
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449032500 250 mL Glass bottle
AC449030010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449032500 Supplier Thermo Scientific Chemicals Supplier No. 449032500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CAS: 111-45-5 | C5H10O2 | 102.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-45-5
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00020607
InChI Key GCYHRYNSUGLLMA-UHFFFAOYSA-N
Synonym 2-allyloxyethanol, 2-allyloxy ethanol, allyl glycol, ethylene glycol monoallyl ether, ethanol, 2-2-propenyloxy, allyl cellosolve, 2-alloxyethanol, usaf do-47, 2-2-propenyloxy ethanol, allyl 2-hydroxyethyl ether
PubChem CID 8116
IUPAC Name 2-prop-2-enoxyethanol
SMILES OCCOCC=C

Specifications

Density 0.9550g/mL
Boiling Point 159.0°C
Flash Point 66°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHCH2OCH2CH2OH
Quantity 250 mL
Specific Gravity 0.955
Solubility Information Solubility in water: soluble
Formula Weight 102.13
Percent Purity 98%
Chemical Name or Material 2-Allyloxyethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 203-871-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.