Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Amino-3-chlorobenzoic acid, 98%
Click to view available options
Quantity:
25 g
100 g
Description
2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Air & Light Sensitive. Store away from air. Store in the dark. Incompatible with light, air and oxidizing agents.
2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.
Solubility
Soluble in water.
Notes
Air & Light Sensitive. Store away from air. Store in the dark. Incompatible with light, air and oxidizing agents.
Specifications
Specifications
| Melting Point | 189°C to 193°C |
| Color | Yellow to Brown |
| Quantity | 25 g |
| Beilstein | 2090431 |
| Sensitivity | Air and light sensitive |
| Solubility Information | Soluble in water. |
| Formula Weight | 171.58 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | 2-Amino-3-chlorobenzoic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 228-996-4 |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction