Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,5-dimethoxybenzoic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
5653-40-7
C9H11NO4
197.19
MFCD00011671
HJVAVGOPTDJYOJ-UHFFFAOYSA-N
4,5-dimethoxyanthranilic acid,6-aminoveratric acid,benzoic acid, 2-amino-4,5-dimethoxy,2-amino-4,5-dimethoxy benzoic acid,2-amino-4,5-dimethoxy-benzoic acid,6-aminoveratricacid,zlchem 238,pubchem3110,acmc-1b03r,ksc269i1n
79736
2-amino-4,5-dimethoxybenzoic acid
COC1=C(C=C(C(=C1)C(=O)O)N)OC
5653-40-7
C9H11NO4
197.19
MFCD00011671
HJVAVGOPTDJYOJ-UHFFFAOYSA-N
4,5-dimethoxyanthranilic acid,6-aminoveratric acid,benzoic acid, 2-amino-4,5-dimethoxy,2-amino-4,5-dimethoxy benzoic acid,2-amino-4,5-dimethoxy-benzoic acid,6-aminoveratricacid,zlchem 238,pubchem3110,acmc-1b03r,ksc269i1n
79736
2-amino-4,5-dimethoxybenzoic acid
COC1=C(C=C(C(=C1)C(=O)O)N)OC
Loss on Drying 1% max.
Color Beige-White or Brown to Gray
Melting Point 156.0°C to 166.0°C
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 197.19
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Amino-4, 5-dimethoxybenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning