Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-chloro-6-methylpyrimidine, 98%, Thermo Scientific™

Catalog No. AC153750050
Click to view available options
:
5g; Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5600-21-5
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Molecular Formula C5H6ClN3
Quantity 5g
MDL Number MFCD00006091
Beilstein 24, 84
Synonym 2-amino-4-chloro-6-methylpyrimidine, am inhibitor, am pesticide, prepn. am, 4-chloro-6-methyl-2-pyrimidinamine, 4-chloro-6-methylpyrimidin-2-ylamine, 2-pyrimidinamine, 4-chloro-6-methyl, am nitrification inhibitor, pyrimidine, 2-amino-4-chloro-6-methyl, 2-amino-6-chloro-4-methylpyrimidine
InChI Key NPTGVVKPLWFPPX-UHFFFAOYSA-N
SMILES CC1=CC(=NC(=N1)N)Cl
IUPAC Name 4-chloro-6-methylpyrimidin-2-amine
Molecular Weight (g/mol) 143.574
PubChem CID 21810
ChEBI CHEBI:58960
Formula Weight 143.58
Percent Purity ≥97.5%
Chemical Name or Material 2-Amino-4-chloro-6-methylpyrimidine, 98%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 227-018-3
RTECSNumber UV6329600
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.