Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-5-chloropyrimidine, 97%, Thermo Scientific™

Catalog No. AC443760010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443760010 1 g
AC443760050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443760010 Supplier Thermo Scientific Chemicals Supplier No. 443760010
Add to Cart

Chemical Identifiers

CAS 5428-89-7
Molecular Formula C4H4ClN3
Molecular Weight (g/mol) 129.55
InChI Key OQZMDDKDHRIGDY-UHFFFAOYSA-N
Synonym 2-amino-5-chloropyrimidine, 2-pyrimidinamine, 5-chloro, 5-chloro-2-pyrimidinamine, pyrimidine, 2-amino-5-chloro, 5-chloropyrimidine-2-ylamine, 5-chlorpyrimidin-2-amin, chloropyrimidinamine, zlchem 634, pubchem9619, acmc-1aya8
PubChem CID 79479
IUPAC Name 5-chloropyrimidin-2-amine
SMILES C1=C(C=NC(=N1)N)Cl

Specifications

Melting Point 235.0°C to 238.0°C
Boiling Point 314.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: slightly soluble.
Formula Weight 129.55
Percent Purity 97%
Chemical Name or Material 2-Amino-5-chloropyrimidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful if swallowed. Causes serious eye damage.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 226-581-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.