Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-6-iodopurine 98.0+%, TCI America™
SDP

Catalog No. A206825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A206825G 25 g
A20685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A206825G Supplier TCI America Supplier No. A206825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19690-23-4
Molecular Formula C5H4IN5
Molecular Weight (g/mol) 261.03
MDL Number MFCD03840496,MFCD08706375
InChI Key CQYPNVKLVHHOSJ-UHFFFAOYSA-N
Synonym 2-amino-6-iodopurine, 6-iodo-9h-purin-2-amine, 2-amino-6-iodo purine, 2-amino-6-iodo-7h-purine, pubchem9213, 6-iodo-2-aminopurine, 2-amino-6-iodo-purine, 6-iodopurine-2-ylamine, acmc-1caa5, amtns010
PubChem CID 6101575
IUPAC Name 6-iodo-7H-purin-2-amine
SMILES NC1=NC(I)=C2NC=NC2=N1

Specifications

Melting Point 240°C
Color Yellow
Quantity 25 g
Formula Weight 261.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-iodopurine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.