Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Benzimidazolylthio)acetic acid, 98%

Catalog No. aaa1122106
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAA1122106

Supplier: Thermo Scientific Chemicals A1122106

Only null left
Add to Cart

CAS: 3042-00-0 | C9H7N2O2S | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3042-00-0
Molecular Formula C9H7N2O2S
Molecular Weight (g/mol) 207.23
MDL Number MFCD00022673
InChI Key UYNVBLJQBCTRKV-UHFFFAOYSA-M
Synonym 2-benzimidazolythio acetic acid, 2-benzimidazolylthio-acetic acid, 2-benzimidazolylthio acetic acid, 1h-benzoimidazol-2-ylsulfanyl acetic acid, 1h-benzimidazol-2-ylthio acetic acid, 2-1h-benzo d imidazol-2-yl thio acetic acid, 2-1h-1,3-benzodiazol-2-ylsulfanyl acetic acid, 1h-benzimidazol-2-ylsulfanyl acetic acid, 1h-1,3-benzodiazol-2-ylsulfanyl acetic acid, 2-benzimidazol-2-ylthioacetic acid
PubChem CID 137805
IUPAC Name 2-(1H-benzimidazol-2-ylsulfanyl)acetic acid
SMILES [O-]C(=O)CSC1=NC2=CC=CC=C2N1

Specifications

Melting Point ∼215°C (decomposition)
Quantity 5 g
Beilstein 11825
Formula Weight 208.24
Percent Purity 98%
Chemical Name or Material (2-Benzimidazolylthio)acetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.