Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 2-BFI hydrochloride
GREENER_CHOICE

Catalog No. 034850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-485-0 50 mg
03-481-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 03-485-0 Supplier Tocris Bioscience™ Supplier No. 0348/50
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, selective I2 ligand. Putative agonist

High affinity I2 ligand (Ki = 9.8 nM). Putative I2 agonist; potentiates morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.

Chemical Identifiers

CAS 89196-95-2
Molecular Formula C11H11ClN2O

Specifications

Quantity 50 mg
Formula Weight 222.67
Purity >98%
Molecular Weight (g/mol) 222.672
Chemical Name or Material 2-BFI hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.