Learn More
2-Bromo-3,3,3-trifluoro-1-propene, 97%
Description
It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as intermediate in production of organ fluorine compounds & as a monomer in fluoropolymer production. It is used as a trifluoromethylated synthetic building block for the synthesis of trifluoromethyl-substituted molecules.
Solubility
Soluble in most organic solvents, immiscible with water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
Specifications
| Refractive Index | 1.353 |
| Quantity | 5 g |
| Solubility Information | Soluble in most organic solvents,immiscible with water. |
| Formula Weight | 174.95 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Bromo-3,3,3-trifluoro-1-propene |
Safety and Handling
| Hazard Category | H335-H336 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P271-P304+P340-P312-P501c |
| TSCA | Yes |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.