Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4,6-dinitroaniline, 97%, Thermo Scientific™

Catalog No. aab2290514
Click to view available options
Quantity:
25 g
100 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1817-73-8
Molecular Formula C6H4BrN3O4
Molecular Weight (g/mol) 262.019
MDL Number MFCD00007146
InChI Key KWMDHCLJYMVBNS-UHFFFAOYSA-N
Synonym 6-bromo-2,4-dinitroaniline, bromo dna, 2,4-dinitro-6-bromoaniline, benzenamine, 2-bromo-4,6-dinitro, 2-bromo-4,6-dinitroaminobenzene, 2,4-dinitro-6-bromanilin, aniline, 2,4-dinitro-6-bromo, aniline, 2-bromo-4,6-dinitro, ccris 820, 2,4-dinitro-6-bromanilin czech
PubChem CID 15752
IUPAC Name 2-bromo-4,6-dinitroaniline
SMILES C1=C(C=C(C(=C1[N+](=O)[O-])N)Br)[N+](=O)[O-]

Specifications

Melting Point 151°C to 153°C
Quantity 25 g
Formula Weight 262.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,6-dinitroaniline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-329-2
TSCA Yes
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.