Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B384625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B384625G 25 g
B38465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B384625G Supplier TCI America Supplier No. B384625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22921-68-2
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
MDL Number MFCD00020214
InChI Key ODHJOROUCITYNF-UHFFFAOYSA-N
Synonym 6-bromo-m-anisic acid, 2-bromo-5-methoxybenzoicacid, benzoic acid, 2-bromo-5-methoxy, 5-methoxy-2-bromobenzoic acid, 2-bromo-5-methoxy-benzoic acid, pubchem2674, acmc-1clk6, ksc205k0l, rarechem al be 1111, 2-bromo-5-methoxy benzoic acid
PubChem CID 89906
IUPAC Name 2-bromo-5-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)Br)C(=O)O

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 25 g
Formula Weight 231.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-methoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.