Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-N-methylaniline, 95%

Catalog No. p-7052652
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55166MD 250 mg
AAH5516603 1 g
AAH5516606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55166MD Supplier Thermo Scientific Chemicals Supplier No. H55166MD
Only null left
Add to Cart

CAS: 6832-87-7 | C7H8BrN | 186.052 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6832-87-7
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD06798064
InChI Key SMVIAQFTVWDWDS-UHFFFAOYSA-N
PubChem CID 334086
IUPAC Name 2-bromo-N-methylaniline
SMILES CNC1=CC=CC=C1Br

Specifications

Color Brown
Density 1.589 g/mL
Boiling Point 107°C to 109°C (12 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.607
Quantity 250 mg
Synonym 2-bromo-n-methylbenzenamine, acmc-20abqa, o-bromo-n-methylaniline, n-methyl-2-bromoaniline, benzenamine,2-bromo-n-methyl, 2-bromo-n-methylaniline, benzenamine, 2-bromo-n-methyl
Formula Weight 186.05
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Bromo-N-methylaniline
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.