Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoacetophenone, 98%

Catalog No. p-71520
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

Catalog No. AC152010250

Supplier: Thermo Scientific Chemicals 152010250

Only null left
Add to Cart

CAS: 70-11-1 | C8H7BrO | 199.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70-11-1
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.05
MDL Number MFCD00000195
InChI Key LIGACIXOYTUXAW-UHFFFAOYSA-N
Synonym 2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo
PubChem CID 6259
ChEBI CHEBI:51846
IUPAC Name 2-bromo-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 46.0°C to 51.0°C
Color Brown-Green to White
Boiling Point 135.0°C (18.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula C6H5COCH2Br
Quantity 25 g
Beilstein 07, 283
Fieser 02,316; 12,381
Merck Index 15, 1409
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chloroform,and ether
Formula Weight 199.05
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material 2-Bromoacetophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 200-724-9
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.