Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromobenzylamine hydrochloride, 97%

Click to view available options
Quantity:
5g
25g
100g
5465-63-4
C7H9BrClN
222.51
MFCD00012853
MDEJOHXOXKDGOU-UHFFFAOYSA-N
2-bromobenzylamine hydrochloride,2-bromophenyl methanamine hydrochloride,o-bromobenzylamine hydrochloride,1-2-bromophenyl methanamine hydrochloride,2-bromobenzylamine hcl,1-2-bromophenyl methylamine hydrochloride,2-bromophenyl methylamine, chloride,benzenemethanamine, 2-bromo-, hydrochloride,pubchem3749,acmc-1aup2
2724202
(2-bromophenyl)methanamine;hydrochloride
C1=CC=C(C(=C1)CN)Br.Cl
5465-63-4
C7H9BrClN
222.51
MFCD00012853
MDEJOHXOXKDGOU-UHFFFAOYSA-N
2-bromobenzylamine hydrochloride,2-bromophenyl methanamine hydrochloride,o-bromobenzylamine hydrochloride,1-2-bromophenyl methanamine hydrochloride,2-bromobenzylamine hcl,1-2-bromophenyl methylamine hydrochloride,2-bromophenyl methylamine, chloride,benzenemethanamine, 2-bromo-, hydrochloride,pubchem3749,acmc-1aup2
2724202
(2-bromophenyl)methanamine;hydrochloride
C1=CC=C(C(=C1)CN)Br.Cl
Applications
It is employed as an intermediate in organic syntheses.

Solubility
Soluble in methanol (25 mg/ml).

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Sensitivity Hygroscopic
Quantity 25g
Melting Point 227°C to 230°C
Beilstein 3913456
Solubility Information Soluble in methanol (25mg/ml).
Formula Weight 222.52
Percent Purity 97%
Chemical Name or Material 2-Bromobenzylamine hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures