Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butoxyethanol, 99%, extra pure

Catalog No. p-3735784
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
25 L
Packaging:
Glass Bottle
Metal drum
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154330010 1 L Glass Bottle
AC154330025 2.5 L Glass Bottle
AC154330100 10 L Plastic drum
AC154330250 25 L Metal drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC154330010 Supplier Thermo Scientific Chemicals Supplier No. 154330010
Only null left
Add to Cart

CAS: 111-76-2 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
PubChem CID 8133
ChEBI CHEBI:63921
SMILES CCCCOCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -70.0°C
Density 0.9010g/mL
Boiling Point 171.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4180 to 1.4200
Linear Formula CH3(CH2)3OCH2CH2OH
Quantity 1 L
Residue after Evaporation 0.02% max.
Beilstein 01, II, 519
Specific Gravity 0.901
Merck Index 15, 1560
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most organic solvents
IUPAC Name 2-butoxyethan-1-ol
Viscosity 5.31 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 99%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material 2-Butoxyethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
EINECSNumber 203-905-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.