Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Butoxyethanol, BAKER™, J.T. Baker™

Catalog No. 02002077 Shop All Avantor Products
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle

Catalog No. 02-002-077

Supplier: Avantor D64807

Only null left
Add to Cart
Add to Cart

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 111-76-2
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00002884
InChI Key POAOYUHQDCAZBD-UHFFFAOYSA-N
Synonym butoxyethanol, butyl glycol, butyl cellosolve, ethylene glycol monobutyl ether, n-butoxyethanol, ethanol, 2-butoxy, ethylene glycol butyl ether, butyl oxitol, glycol butyl ether, glycol ether eb
PubChem CID 8133
ChEBI CHEBI:63921
SMILES CCCCOCCO

Specifications

Flash Point 62.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500 mL
Specific Gravity 1L = 0.90kg
IUPAC Name 2-butoxyethanol
Formula Weight 118.18
Chemical Name or Material 2-Butoxyethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.