Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc nitrate hexahydrate, 98%, extra pure

Catalog No. AC211660051
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle

CAS: 10196-18-6 | H12N2O12Zn | 297.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 36.0°C
Color White
Physical Form Adhering Crystals
Quantity 5 kg
Assay Percent Range 98%
Linear Formula Zn(NO3)2·6H2O
Merck Index 14,10144
Solubility Information Solubility in water: 1800g/L (20°C). Other solubilities: freely soluble in alcohol
IUPAC Name zinc(2+) hexahydrate dinitrate
Formula Weight 297.46
Percent Purity 98%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Zinc nitrate hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye pr

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.