Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorophenethylamine, 97%, Thermo Scientific™

Catalog No. p-87420
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC350640010

Supplier: Thermo Scientific Chemicals 350640010

Only null left
Add to Cart

Chemical Identifiers

CAS 149488-93-7
Molecular Formula C8H9ClFN
Molecular Weight (g/mol) 173.62
MDL Number MFCD00052682
InChI Key NNEMMAAHBKHXLP-UHFFFAOYSA-N
PubChem CID 2734094
IUPAC Name 2-(2-chloro-6-fluorophenyl)ethanamine
SMILES C1=CC(=C(C(=C1)Cl)CCN)F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Pink-Red
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5285 to 1.5305
Quantity 1 g
Synonym 2-chloro-6-fluorophenethylamine, 2-2-chloro-6-fluorophenyl ethanamine, 2-2-chloro-6-fluorophenyl ethylamine, benzeneethanamine,2-chloro-6-fluoro, 2-2-chloro-6-fluoro-phenyl-ethylamine, 2-2-chloro-6-fluorophenyl ethan-1-amine, 2-6-chloro-2-fluorophenyl ethylamine, acmc-1c8qt, benzeneethanamine, 2-chloro-6-fluoro
Formula Weight 173.62
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-fluorophenethylamine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.