Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-chloro-N,N-dimethylacetamide, 97%

Catalog No. p-4228504
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle

CAS: 2675-89-0 | C4H8ClNO | 121.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2675-89-0
Molecular Formula C4H8ClNO
Molecular Weight (g/mol) 121.57
InChI Key XBPPLECAZBTMMK-UHFFFAOYSA-N
Synonym n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide, acetamide, 2-chloro-n,n-dimethyl, 2-chloro-n, n-dimethylacetamide, acmc-209gsx, 2-chlorodimethylacetamide, 2-chloro-dimethylacetamide, 2-chlorodimethyl acetamide, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide
PubChem CID 75886
IUPAC Name 2-chloro-N,N-dimethylacetamide
SMILES CN(C)C(=O)CCl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 15°C to 16°C
Color Colorless to Yellow
Density 1.1820g/mL
Boiling Point 98°C to 100°C (11.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CON(CH3)2
Quantity 25 mL
Specific Gravity 1.182
Solubility Information Solubility in water: in water:(20°C) soluble.,in ethanol:(20°C) soluble
Formula Weight 121.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chloro-N, N-dimethylacetamide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-224-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.