Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Benzenesulfonate 98.0+%, TCI America™
SDP

Catalog No. C223025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C223025G 25 g
C22305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C223025G Supplier TCI America Supplier No. C223025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16670-48-7
Molecular Formula C8H9ClO3S
Molecular Weight (g/mol) 220.667
InChI Key CIIGWOXXOUVEAD-UHFFFAOYSA-N
Synonym Benzenesulfonic Acid 2-Chloroethyl Ester
PubChem CID 13634485
IUPAC Name 2-chloroethyl benzenesulfonate
SMILES C1=CC=C(C=C1)S(=O)(=O)OCCCl

Specifications

Color Yellow
Boiling Point 188°C
Quantity 25 g
Formula Weight 220.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Benzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.