Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl Chloromethyl Ether 98.0+%, TCI America™
SDP

Catalog No. C140525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C140525G 25 g
C14055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C140525G Supplier TCI America Supplier No. C140525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1462-33-5
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.98
MDL Number MFCD00191406
InChI Key LUTWEKBTDWRTSE-UHFFFAOYSA-N
Synonym 2-(Chloromethoxy)ethyl Chloride
PubChem CID 10290810
IUPAC Name 1-chloro-2-(chloromethoxy)ethane
SMILES ClCCOCCl

Specifications

Color Colorless
Boiling Point 147°C
Quantity 25 g
UN Number 2929
Formula Weight 128.98
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Chloromethyl Ether
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.