Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloroethyl ethyl ether, 98+%

Catalog No. p-7027507
Click to view available options
Quantity:
100 g

CAS: 628-34-2 | C4H9ClO | 108.57 g/mol

It is used as a pharmaceutical intermediate. Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2 illuminated with UV light and maintained at 25 or 80°C in air was found to suffer progressive oxidation to yield ethylene, chloroethylene , ethanol, acetaldehyde, chloroacetaldehyde, diethyl disulfide, 2-chloroethyl ethyl disulfide and bis(2-chloroethyl) disulfide as the main primary intermediates, and water (H2O), carbon dioxide (CO2), sulfur dioxide (SO2), surface sulfate ions (SO42-), and hydrogen chloride (HCl) as the final products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. Photocatalytic oxidation of gaseous 2-chloroethyl ethyl sulfide over TiO2 illuminated with UV light and maintained at 25 or 80°C in air was found to suffer progressive oxidation to yield ethylene, chloroethylene , ethanol, acetaldehyde, chloroacetaldehyde, diethyl disulfide, 2-chloroethyl ethyl disulfide and bis(2-chloroethyl) disulfide as the main primary intermediates, and water (H2O), carbon dioxide (CO2), sulfur dioxide (SO2), surface sulfate ions (SO42-), and hydrogen chloride (HCl) as the final products.

Solubility
Miscible with water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-34-2
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00000974
InChI Key GPTVQTPMFOLLOA-UHFFFAOYSA-N
Synonym 2-chloroethyl ethyl ether, 2-ethoxyethyl chloride, 2-chloroethylethylether, 2-chloroethoxyethane, 1-chloro-2-ethoxy-ethane, ether, 2-chloroethyl ethyl, beta-chloroethyl ethyl ether, ethyl beta-chloroethyl ether, ethane, 1-chloro-2-ethoxy, ccris 9095
PubChem CID 12341
IUPAC Name 1-chloro-2-ethoxyethane
SMILES CCOCCCl

Specifications

Melting Point -60°C
Density 0.99
Boiling Point 107°C to 108°C
Flash Point 6°C (43°F)
Refractive Index 1.412
Quantity 100 g
UN Number UN3271
Solubility Information Miscible with water.
Formula Weight 108.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloroethyl ethyl ether
Show More Show Less
Hazard Category H225-H302+H332-H315-H318-H335
Hazard Statement GHS H Statement
H225-H331-H315-H319-H335
Highly flammable liquid and vapour.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 211-037-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.