Alfa Aesar™ 2-Chloroethyl p-toluenesulfonate, 97%

Manufacturer: Alfa Aesar™ A1186336

Quantity 500g

 View more versions of this product

Catalog No. AAA1186336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80-41-1
    Molecular Formula C9H11ClO3S
    Formula Weight 234.7
    MDL Number MFCD00000970
    Beilstein 1968816
    Synonym p-Toluenesulfonic acid 2-chloroethyl ester
    Chemical Name or Material 2-Chloroethyl p-toluenesulfonate

    Specifications

    Formula Weight 234.7
    TSCA TSCA
    Synonym p-Toluenesulfonic acid 2-chloroethyl ester
    Health Hazard 1 Warning
    Molecular Formula C9H11ClO3S
    CAS 80-41-1
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Recommended Storage 2°C to 8°C
    RTECS Number XT6475000
    MDL Number MFCD00000970
    Beilstein 1968816
    Health Hazard 3 GHS P Statement
    P280f
    Wear protective clothing.
    Flash Point >110°C (230°F)
    Boiling Point 158° to 159°C (0.5mmHg)
    Quantity 500g
    Chemical Name or Material 2-Chloroethyl p-toluenesulfonate
    Assay 97%

    GHS H Statement
    H302
    Harmful if swallowed.

    2°C to 8°C

    GHS P Statement
    P280f
    Wear protective clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit