Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl bromide, 1M solution in THF, AcroSeal™

Catalog No. AC428778000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle

CAS: 593-60-2 | C2H3Br | 106.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Vinyl bromide
Name Note 1M solution in THF
CAS 593-60-2, 109-99-9
Density 0.9270g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C2H3Br
Linear Formula H2C=CHBr
Quantity 800 mL
MDL Number MFCD00000183
Specific Gravity 0.927
Synonym vinyl bromide, bromoethylene, ethene, bromo, monobromoethylene, vinylbromid, bromure de vinyle, vinylbromide, polyvinyl bromide, ethylene, bromo, polyvinylbromide
InChI Key INLLPKCGLOXCIV-UHFFFAOYSA-N
SMILES BrC=C
IUPAC Name bromoethene
Molecular Weight (g/mol) 106.95
PubChem CID 11641
ChEBI CHEBI:51311
Formula Weight 106.95
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.