Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Chloromethyl)benzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. c31525g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g

Chemical Identifiers

CAS 42908-86-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00152348
InChI Key TXZFBHYDQGYOIT-UHFFFAOYSA-N
PubChem CID 3016395
IUPAC Name 2-(chloromethyl)benzoyl chloride
SMILES ClCC1=CC=CC=C1C(Cl)=O

Specifications

Melting Point 17°C
Color Yellow
Boiling Point 265°C
Quantity 5 g
UN Number 3265
Formula Weight 189.04
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(Chloromethyl)benzoyl Chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.