Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cadmium sulfate, 8/3-hydrate, ACS reagent, crystals

Catalog No. AC403765000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle

CAS: 7790-84-3 | Cd3O12S3·8H2O | 769.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
Molecular Formula Cd3O12S3·8H2O
Molecular Weight (g/mol) 769.51
MDL Number MFCD02253000

Specifications

Melting Point 41.0°C
Color White
Physical Form Crystals
Quantity 500 g
Linear Formula 3CdSO4·8H2O
Merck Index 15,1632
Solubility Information Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol, acids, acetone and ammonia
Formula Weight 769.51
Percent Purity 98.0 to 102.0%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Cadmium sulfate, 8/3-hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
May cause genetic defects.
May damage fertility.
May damage the unborn child.
Fatal if inhaled.
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Immediately call a POISON CENTER or doctor/physician.
Do not breathe dust

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.