Learn More
2-Chloropropionyl chloride, 96%
Description
2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chloropropionyl chloride is used as a reagent in the chemical surface fictionalization of parylene C. It is used in the synthesis of (±)-mintlactone, in the preparation of a poly(ethylene glycol) macro initiator & chlorine modified TiO2 nanoparticles, the ATRP initiators.
Solubility
Reacts with water.
Notes
Store away from oxidizing agents, active metals, bases, water/ moisture.

Specifications
Specifications
Density | 1.308 |
Boiling Point | 109°C to 111°C |
Flash Point | 31°C (88°F) |
Refractive Index | 1.44 |
Quantity | 100 g |
UN Number | UN2920 |
Solubility Information | Reacts with water. |
Formula Weight | 126.97 |
Percent Purity | 96% |
Chemical Name or Material | 2-Chloropropionyl chloride |
Safety and Handling
Hazard Category | H226-H302-H314-H335 |
Hazard Statement | GHS H Statement H301-H314-H318-H226 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S. |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.