Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 2-Deoxy-D-glucose
GREENER_CHOICESDP

Catalog No. 451550 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-155-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-155-0 Supplier Tocris Bioscience™ Supplier No. 4515/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Non-metabolizable glucose analog; hexokinase substrate

2-Deoxy-D-glucose is a non-metabolizable glucose analog. Inhibits phosphorylation of glucose by hexokinase; causes depletion of cellular ATP. Also inhibits phosphoglucose isomerase (PGI) competitively. Causes cell cycle inhibition and cell death in in vitro models of hypoxia; blocks tumor cell growth in animal models. Also shown to induce the unfolded protein response (UPR). 2-Deoxy-D-glucose disrupts N-glycan expression on the surface of pancreatic adenocarcinoma cells. 2-Deoxy-D-glucose enhances chimeric antigen receptor T cell activity in different xenograft mouse models of pancreatic adenocarcinoma cells.

Chemical Identifiers

CAS 154-17-6
Molecular Formula C6H12O5

Specifications

Quantity 50 mg
Formula Weight 164.16
Purity >98%
Molecular Weight (g/mol) 164.16
Chemical Name or Material 2-Deoxy-D-glucose
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.