Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Diethylamino)ethanol, 99%, Thermo Scientific Chemicals

Catalog No. AAA162740J
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL

Diethylethanolamine can be used as a precursor chemical to procaine. It is used as a corrosion inhibitor in steam and condensate lines by neutralizing carbonic acid and scavenging oxygen. it is used for the synthesis of drugs in the pharmaceutical industry and as a catalyst for the synthesis of polymers in the chemical industry. It is also used as a pH stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethylethanolamine can be used as a precursor chemical to procaine. It is used as a corrosion inhibitor in steam and condensate lines by neutralizing carbonic acid and scavenging oxygen. it is used for the synthesis of drugs in the pharmaceutical industry and as a catalyst for the synthesis of polymers in the chemical industry. It is also used as a pH stabilizer.

Solubility
It is miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic , Avoid moisture. Keep away from strong oxidizing agents, strong acids, Copper, Zinc, Iron, Do not store near acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-37-8
Molecular Formula C6H15NO
MDL Number MFCD00002850
InChI Key BFSVOASYOCHEOV-UHFFFAOYSA-N
Synonym 2-diethylamino ethanol, n,n-diethylethanolamine, diethylaminoethanol, deae, diethylethanolamine, diethylamino ethanol, ethanol, 2-diethylamino, n,n-diethyl-2-aminoethanol, 2-hydroxyethyl diethylamine, diethyl 2-hydroxyethyl amine
PubChem CID 7497
ChEBI CHEBI:52153
IUPAC Name 2-(diethylamino)ethanol
SMILES CCN(CC)CCO

Specifications

Melting Point -70°C
pH 11.5
Boiling Point 161°C to 163°C
Flash Point 52°C (125°F)
Odor Amine-like
Linear Formula (CH3CH2)2NCH2CH2OH
Quantity 5000 mL
UN Number UN2686
Beilstein 741863
Merck Index 14,3112
Molecular Weight (g/mol) 117.192
Formula Weight 117.19
Percent Purity 99%
Chemical Name or Material 2-(Diethylamino)ethanol
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P280-P262-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes, on skin, or on clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: 2-DIETHYLAMINOETHANOL
RTECSNumber KK5075000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.