Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Ethoxyethyl acetate, 98+%, pure, Thermo Scientific™
Click to view available options
:
5mL; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
CAS | 111-15-9 |
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Density | 0.9700g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Glass Bottle |
Molecular Formula | C6H12O3 |
Refractive Index | 1.4040 to 1.4060 (20°C, 589nm) |
Linear Formula | CH3CO2CH2CH2OC2H5 |
Show More |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement: Harmful in contact with skin. May damage fertility. May damage the unborn child. Harmful if swallowed. Harmful if inhaled. Flammable liquid and vapor. |
Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 203-839-2 |
RTECSNumber | KK8225000 |
TSCA | TSCA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction