Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxypyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. E11155G
Click to view available options
Quantity:
1 g
5 g

Specifications

CAS 612845-44-0
Melting Point 134°C
Color White
Molecular Formula C7H10BNO3
MDL Number MFCD06201032
Quantity 5 g
Synonym 6-ethoxypyridin-3-yl boronic acid, 2-ethoxy-5-pyridineboronic acid, 2-ethoxypyridine-5-boronic acid, 6-ethoxypyridine-3-boronic acid, 6-ethoxy-3-pyridinylboronic acid
InChI Key UONCERAQKBPLML-UHFFFAOYSA-N
SMILES B(C1=CN=C(C=C1)OCC)(O)O
IUPAC Name (6-ethoxypyridin-3-yl)boronic acid
Molecular Weight (g/mol) 166.971
PubChem CID 12012237
Formula Weight 166.97
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.