Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethylimidazole, 98%

Catalog No. p-7027382
Click to view available options
Quantity:
100 g
500 g

CAS: 1072-62-4 | C5H8N2 | 96.13 g/mol

2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylimidazole is used as internal standard in the quantification of 4-(5-)methylimidazole (4MI) in coffee by GC-MS method. It is a versatile intermediate used as a building block for active ingredients as well as in epoxy curing.

Solubility
Soluble in water (617 g/L at 20°C)

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-62-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005192
InChI Key PQAMFDRRWURCFQ-UHFFFAOYSA-N
Synonym 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PubChem CID 66130
IUPAC Name 2-ethyl-1H-imidazole
SMILES CCC1=NC=CN1

Specifications

Melting Point 78°C to 86°C
Boiling Point 268°C
Flash Point 159°C (318°F)
Quantity 100 g
Beilstein 106381
Solubility Information Soluble in water (617g/L at 20°C)
Formula Weight 96.13
Percent Purity 98%
Chemical Name or Material 2-Ethylimidazole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P234-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P390-P501c
EINECSNumber 214-011-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.